Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Wire baskets

140×140×140 mm
  215-7414
Wire baskets
  215-7414
Wire baskets
Wire baskets

140×140×140 mm

 
171,00 €
   
1 * 1 ST
 

Stainless steel.

  • Spot welded mesh 8×8×1 mm
  • Electrolytically polished
  • Reinforced edges

These baskets can be used for transport, storage, cleaning and sterilisation of laboratory utensils.

 
  • Seq:
    04
  • W×D×H:
    140×140×140 mm