Replacement caps
SVL 20
Artikelnr. 215-2044
Anbieter: QVF LABORTECHNIK
Jump to:
Product Details & Documents
Pyrex®, phenoplast, black.
- Excellent chemical resistance, excellent temperature resistance
- Screw caps with PTFE-coated seal
- Replacement seals are available as an option
Replacement closures for reagent and sample glasses as well as bottles with SVL threads.
Specifications
- Thread:SVL 20