Storage Boxes for Vials
10×10, Blue, 140×153×55 mm
Catalog # NEOL2-1938
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Product Details & Documents
Storage boxes, made of PP, coloured, for HPLC vials, cryotubes or 1,5 and 2,0 ml microcentrifuge tubes with the height of 46 mm and up to 12 mm diameter.
- Stackable
- Transparent, tight sealing and hinged lid
- Also usable as cryobox
- Autoclavable
Specifications
- Compartments:10×10
- W×D×H ext.:140×153×55 mm
- Colour:Blue
- Seq:04
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



