Orders LinkContactUsLinkComponent
Evaporating flasks
Evaporating flasks
100 ml, 29/42 NS, uncoated
Catalog # BUCH008737
Supplier:   BUCHI
Evaporating flasks
Catalog # BUCH008737
Supplier:  BUCHI
Evaporating flasks
Evaporating flasks
100 ml, 29/42 NS, uncoated
Price
144,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For Rotavapor® rotary evaporators. Centre neck. Pear-shaped with ground joint socket or EL-flange.
Specifications
  • Socket size:
    29/42 NS
  • Type:
    uncoated
  • Capacity:
    100 ml
  • Seq:
    06