Orders LinkContactUsLinkComponent
Drying flasks
Drying flasks
2000 ml, 29/32 NS
Catalog # 531-1983
Supplier:   BUCHI
Drying flasks
Catalog # 531-1983
Supplier:  BUCHI
Drying flasks
Drying flasks
2000 ml, 29/32 NS
Price
230,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Made of Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For rotary evaporators Rotavapor R-210/215. Pear shape with NS 29/32 neck. Glassware A, V, C.
Specifications
  • Socket size:
    29/32 NS
  • Capacity:
    2000 ml