Orders LinkContactUsLinkComponent
Adapters, bends
29/32 NS
  271-2203
Adapters, bends
  271-2203
Adapters, bends
Adapters, bends
29/32 NS
36,90 €
1 * 1 ST
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Inclined at an angle of 90°. With 2 standard ground cones.
  • Cone size:
    29/32 NS