Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer solutions

Buffer solutions

Type L6874, 6,87, ±0,010, 25 °C, 250 mL
Catalog # 662-6182
Supplier:   SI Analytics
Buffer solutions
Catalog # 662-6182
Supplier:  SI Analytics
CAS Number:  
Buffer solutions
Buffer solutions

Type L6874, 6,87, ±0,010, 25 °C, 250 mL

CAS Number:  
Price
87,00 €
Log in for availability
1 * 250 mL
Product Details & Documents

Buffer solutions intended for extremely precise calibration.

  • Highest measurement reliability
  • No preservative agents
  • Hermetically sealed in the glass ampoule
  • Sterilised with hot steam

Certifications: According to DIN 19 266.

Specifications

Specifications

  • Pk:
    250 mL
  • Reference temperature:
    25 °C
  • Description:
    Type L6874
  • pH accuracy:
    ±0,010
  • pH value:
    6,87

Specifications