Orders LinkContactUsLinkComponent
Cables for Electrodes
Plug cable combination, VP 6-19/05 (VP 6-pole electrode socket, 5 m cable KA 19 (2 coaxial and 3 cords) upto 130 °C, free end), 5 m
Catalog # 662-2258
Supplier:   SI Analytics
Cables for Electrodes
Catalog # 662-2258
Supplier:  SI Analytics
Cables for Electrodes
Cables for Electrodes
Plug cable combination, VP 6-19/05 (VP 6-pole electrode socket, 5 m cable KA 19 (2 coaxial and 3 cords) upto 130 °C, free end), 5 m
Price
272,00 €
Log in for availability
1 * 1 ST
Specifications
  • Description:
    Plug cable combination, VP 6-19/05 (VP 6-pole electrode socket, 5 m cable KA 19 (2 coaxial and 3 cords) upto 130 °C, free end)
  • Length:
    5 m
  • Seq:
    28