Order Entry
Austria
Orders LinkContactUsLinkComponent
Cables for Electrodes

Cables for Electrodes

Plug cable combination, K 10 X HT (coaxial electrode plug, 10 m coaxial cable, 135 °C, free end), 10 m
Catalog # 662-2206
Supplier:   SI Analytics
Cables for Electrodes
Catalog # 662-2206
Supplier:  SI Analytics
Cables for Electrodes
Cables for Electrodes

Plug cable combination, K 10 X HT (coaxial electrode plug, 10 m coaxial cable, 135 °C, free end), 10 m

Price
185,00 €
Log in for availability
1 * 1 ST
Specifications
  • Description:
    Plug cable combination, K 10 X HT (coaxial electrode plug, 10 m coaxial cable, 135 °C, free end)
  • Length:
    10 m
  • Seq:
    18
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.