Orders LinkContactUsLinkComponent
Electrode Cables
Electrode Cables
Without connector (free end) for custom installation, 1 m
Catalog # 662-6118
Supplier:   SI Analytics
Electrode Cables
Catalog # 662-6118
Supplier:  SI Analytics
Electrode Cables
Electrode Cables
Without connector (free end) for custom installation, 1 m
Price
42,40 €
Log in for availability
1 * 1 ST
Product Details & Documents
Electrode cables with a range of connectors to fit most pH meters.
Specifications
  • Description:
    Without connector (free end) for custom installation
  • Length:
    1 m
  • Seq:
    01