Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Electrode Cables

Electrode cable with 2 mm pole connector, 1 m
  662-6124
Electrode Cables
  662-6124
Electrode Cables
Electrode Cables

Electrode cable with 2 mm pole connector, 1 m

 
39,80 €
   
1 * 1 ST
 
 
  • Description:
    Electrode cable with 2 mm pole connector
  • Length:
    1 m
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.