J.T.Baker® BAKER ANALYZED®, Buffer Solutions
4,00, ±0,02, Citrate, 20 °C, Plastic bottle, 1 L
Supplier: Avantor
Jump to:
Product Details & Documents
Ready-to-use pH buffers are manufactured in large lots to stringent specifications and utilise quality control procedures to reduce lot-to-lot variability. You can eliminate the time and expense of preparation and standardisation of solutions, helping streamline high-volume analytical work in applications such as quality control.
Contain 0,01% Sodium benzoate.
Certifications: This buffer is traceable to NIST Standard reference buffer.
Specifications
Specifications
- Pack type:Plastic bottle
- pH buffer type:Citrate
- Pk:1 L
- Reference temperature:20 °C
- pH accuracy:±0,02
- pH value:4,00
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



