Order Entry
Austria
Orders LinkContactUsLinkComponent
pH-Fix pH Indicator Test Strips

pH-Fix pH Indicator Test Strips

pH-Fix, 100 test strips with colour scale, 7,5 - 9,5, Classic flat box , 66×10×90 mm, 34,58 g
Catalog # 662-0236
Supplier:   MACHEREY-NAGEL
pH-Fix pH Indicator Test Strips
Catalog # 662-0236
Supplier:  MACHEREY-NAGEL
pH-Fix pH Indicator Test Strips
pH-Fix pH Indicator Test Strips

pH-Fix, 100 test strips with colour scale, 7,5 - 9,5, Classic flat box , 66×10×90 mm, 34,58 g

Price
16,10 €
Log in for availability
1 * 100 ST
Product Details & Documents

pH-Fix indicator test strips allow fast pH testing directly at the point of interest, and are suitable for untrained users as well as analytical professionals.

  • No accessories needed - always ready to use
  • Brilliant colour chart for precise readings
  • Long handle for sufficient hand - sample - distance
  • Non bleeding - no contamination of sample solutions

The indicator dyes used for pH-Fix test strips are chemically bound to the test pads to prevent bleeding and, therefore, contamination of the sample. Instructions for use according to IVD Directive 98/79/EC - The instructions for use according to IVD Directive 98/79/EC can be requested by providing the article number and LOT number at the email address [email protected].

Caution: No hazardous material.

Specifications
  • pH range:
    7,5 - 9,5
  • Description:
    pH-Fix, 100 test strips with colour scale
  • Seq:
    16
  • Pack type:
    Classic flat box , 66×10×90 mm, 34,58 g
  • Cat. No.:
    662-0236
  • Shelf life:
    4 Year(s)