Order Entry
Austria
Orders LinkContactUsLinkComponent
Automatic burettes, Schilling pattern, PMMA, class B

Automatic burettes, Schilling pattern, PMMA, class B

25 ml, 0,1 ml, 0,04 ± ml
Catalog # 661-2405
Supplier:   witeg Labortechnik
Automatic burettes, Schilling pattern, PMMA, class B
Catalog # 661-2405
Supplier:  witeg Labortechnik
Automatic burettes, Schilling pattern, PMMA, class B
Automatic burettes, Schilling pattern, PMMA, class B

25 ml, 0,1 ml, 0,04 ± ml

Price
107,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Automatic burettes, class DIN-B, short line graduation, clear plastic (PMMA).

  • Push button enabling a precise titration
  • Complete with polyethylene reservoir
  • Included with a stable base
  • Automatic zero point

Liquid delivery time 30 to 60 seconds, without waiting time.

Specifications
  • Capacity:
    25 ml
  • Seq:
    01
  • Division:
    0,1 ml
  • Tolerance:
    0,04 ± ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.