Order Entry
Austria
Orders LinkContactUsLinkComponent
Tissue processing cassettes, polyacetylene

Tissue processing cassettes, polyacetylene

Yellow
Catalog # 631-9644
Supplier:   DELTALAB
Tissue processing cassettes, polyacetylene
Catalog # 631-9644
Supplier:  DELTALAB
Tissue processing cassettes, polyacetylene

Yellow

Price
140,00 €
Einloggen für Verfügbarkeit
1 * 1.000 ST
Product Details & Documents

These cassettes are made of polyacetyle.

  • With 34 round holes of 2 mm
  • External dimensions: 40×28×6 mm
  • Can be written on

They are resistant to the solvents used in histological processes.

Documents

Spezifikationen
  • Colour:
    Yellow
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.