Order Entry
Austria
Orders LinkContactUsLinkComponent
Tissue embedding cassettes, UNI type

Tissue embedding cassettes, UNI type

Green
Catalog # 631-9614
Supplier:   KARTELL
Tissue embedding cassettes, UNI type
Catalog # 631-9614
Supplier:  KARTELL
Tissue embedding cassettes, UNI type
Tissue embedding cassettes, UNI type

Green

Price
95,90 €
Log in for availability
1 * 500 ST
Product Details & Documents

Tissue embedding cassettes for paraffin inclusions, Universal type, provided with clip on closure lid that can easily be removed. They are available in five colours. Dimensions: 28×40×6,8 mm.

Specifications
  • Colour:
    Green
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Tissue embedding sponge, blue
    Tissue embedding sponge, blue
    KART2922-04
    Log in for availability
    1 * 500 ST - 29,40 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.