Orders LinkContactUsLinkComponent
Staining jars
Staining dishes acc. to Hellendahl, for up to 16 slides
  631-9310
Staining jars
  631-9310
Staining jars
Staining jars
Staining dishes acc. to Hellendahl, for up to 16 slides
29,10 €
1 * 1 ST
Soda-lime glass.
  • Three versions
  • For 16 or 10 slides
  • Acc. to Hellendahl or Schiefferdecker
Suitable for standard microscope slides (76×26 mm).
  • Description:
    Staining dishes acc. to Hellendahl, for up to 16 slides