Inoculating loops, reusable
Loops, length 50 mm, Ø 4,0 mm
Catalog # 631-7133
Supplier: RSG
Springe zu:
Product Details & Documents
These NiCr 8020 loops are available in a range of sizes and lengths with internal diameter of 1,5, 2,5 or 4 mm. They are heat resistant and can be flame sterilised. Diameter of wire: 0,6 mm. Loop holder available separately.
Documents
RSGA268.001_TDS_EN
Datasheet
Spezifikationen
- Description:Loops, length 50 mm, Ø 4,0 mm



