Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Baumé hydrometers

30 - 40 °Bé, 0,1, 280 mm
  614-0539
 :   Amarell
Baumé hydrometers
  614-0539
 :  Amarell
Baumé hydrometers
Baumé hydrometers

30 - 40 °Bé, 0,1, 280 mm

 
32,30 €
   
1 * 1 ST
 

General purpose Baumé hydrometers with or without thermometer.

  • Reference temperature of 15 °C
  • Paper scale with visible print
  • Shot weighted body

Colourless round capillary with red or blue filling.

 
  • Length:
    280 mm
  • Seq:
    19
  • Division:
    0,1
  • Range:
    30 - 40 °Bé
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.