Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Graduated pipettes, with piston, Class B

5 ml, 0,05 ml, 0,04 ± ml, Black
Catalog # 613-2171
Supplier:   witeg Labortechnik
Graduated pipettes, with piston, Class B
Catalog # 613-2171
Supplier:  witeg Labortechnik
Graduated pipettes, with piston, Class B
Graduated pipettes, with piston, Class B

5 ml, 0,05 ml, 0,04 ± ml, Black

Price
15,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

Glass, class DIN-B

  • With ISO-COLOR-Code
  • With interchangeable piston and tempered tips
  • With amber stain DIFFICO graduation and inscriptions
  • Short line graduation
  • Complete delivery
Specifications
  • Colour code:
    Black
  • Capacity:
    5 ml
  • Division:
    0,05 ml
  • Tolerance:
    0,04 ± ml