Orders LinkContactUsLinkComponent
Pipette controller, Pi-Pump
Pipette controller, Pi-Pump
Pipette controller Pi-Pump, red, for 25 ml pipettes
Catalog # 612-1927
Supplier:   KARTELL
Pipette controller, Pi-Pump
Catalog # 612-1927
Supplier:  KARTELL
Pipette controller, Pi-Pump
Pipette controller, Pi-Pump
Pipette controller Pi-Pump, red, for 25 ml pipettes
Price
24,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
Pipetting aid for all plastic and glass pipettes.
  • Suction and dispensing achieved by rotating the wheel
  • Liquid evacuated by pressing the lever
Specifications
  • Description:
    Pipette controller Pi-Pump, red, for 25 ml pipettes