Refrigerator kiu1444total capacity 137l fridge 120l freezer 27l class a energy rating height 870 x width 600 x depth 570 mm freezer on top premium compact built under refrigerator 1 * 1 items
Catalog # 472-2278
Supplier: Liebherr
Frequently Bought Together