Order Entry
Austria
Orders LinkContactUsLinkComponent
Bath fluids

Bath fluids

Therm 160, 60...160 °C, >273 °C, >263 °C, Brown, 20 l
Catalog # 461-2873
Supplier:   LAUDA
Bath fluids
Catalog # 461-2873
Supplier:  LAUDA
CAS Number:  
Bath fluids
Bath fluids

Therm 160, 60...160 °C, >273 °C, >263 °C, Brown, 20 l

CAS Number:  
Price
1 130,00 €
Log in for availability
1 * 20 L
Product Details & Documents

Allow highly accurate thermostating, even at extreme temperatures.

  • Simple and safe handling, suitable for long term operation
  • Optimise thermostat life

* Denotes Silicone oil
** Only suitable for closed circuits

Specifications

Specifications

  • Pk:
    20 l
  • Colour:
    Brown
  • Fire point:
    >273 °C
  • Flash point:
    >263 °C
  • Temperature range:
    60...160 °C
  • Type:
    Therm 160

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.