Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Bath fluids

Aqua 90, 5...90 °C, − °C, − °C, Colourless, 5 l
Catalog # 461-2858
Supplier:   LAUDA
Bath fluids
Catalog # 461-2858
Supplier:  LAUDA
CAS Number:  
Bath fluids
Bath fluids

Aqua 90, 5...90 °C, − °C, − °C, Colourless, 5 l

CAS Number:  
Price
53,40 €
Log in for availability
1 * 5 L
Product Details & Documents

Allow highly accurate thermostating, even at extreme temperatures.

  • Simple and safe handling, suitable for long term operation
  • Optimise thermostat life

* Denotes Silicone oil
** Only suitable for closed circuits

Specifications

Specifications

  • Pk:
    5 l
  • Colour:
    Colourless
  • Fire point:
    − °C
  • Flash point:
    − °C
  • Temperature range:
    5...90 °C
  • Type:
    Aqua 90

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.