Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Bath fluids, Haake

Sil 100, –75...+75 °C, >100 °C, 57 °C, 3 cP, Colourless, 10 L
Catalog # 461-0011
Supplier:   THERMO ELECTRON LED
Bath fluids, Haake
Catalog # 461-0011
CAS Number:  
Bath fluids, Haake

Sil 100, –75...+75 °C, >100 °C, 57 °C, 3 cP, Colourless, 10 L

CAS Number:  
Price
1 220,00 €
Log in for availability
1 * 10 L
Product Details & Documents

These carefully selected and proven heat transfer liquids have been designed to reduce health hazards and minimise unpleasant odours.

  • Silicone oils carry a low risk of inflammation, do not give off unpleasant odours and have a long service life
  • Low corrosive tendency

The oils are not suitable for contact with silicone tubing.

Specifications

Specifications

  • Pk:
    10 L
  • Colour:
    Colourless
  • Fire point:
    >100 °C
  • Flash point:
    57 °C
  • Temperature range:
    –75...+75 °C
  • Type:
    Sil 100
  • Viscosity:
    3 cP

Specifications