Order Entry
Austria
Orders LinkContactUsLinkComponent
Pestles

Pestles

42 mm, 175 mm, 410-9112
Catalog # 410-9129
Pestles
Catalog # 410-9129
Pestles
Pestles

42 mm, 175 mm, 410-9112

Price
55,10 €
Log in for availability
1 * 1 ST
Product Details & Documents

Porcelain with glazed handle, glazed or unglazed grinding surface.

Certifications: Manufactured to DIN 12906.

Specifications
  • For:
    410-9112
  • Head Ø:
    42 mm
  • Length:
    175 mm
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.