Orders LinkContactUsLinkComponent
Microplates, 96-well
Microplates, 96-well
U-bottom, clear, sterile, 40 - 280 µl, 96, +, 323 µl
Catalog # 391-3311
Supplier:   Greiner Bio-One
Microplates, 96-well
Catalog # 391-3311
Supplier:  Greiner Bio-One
Microplates, 96-well
Microplates, 96-well
U-bottom, clear, sterile, 40 - 280 µl, 96, +, 323 µl
Price
243,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
PS, without lid. A large number of different 96-well microplates are available in a wide variety of surface treatments with a choice of well profile, and with either solid or µClear® bottom. All plates are free of DNase, RNase and human DNA, and are non-pyrogenic.
Specifications
  • No. of wells:
    96
  • Recommended working volume:
    40 - 280 µl
  • Sterile:
    +
  • Description:
    U-bottom, clear, sterile
  • Seq:
    04
  • Total volume:
    323 µl