Orders LinkContactUsLinkComponent
QualiSampler sets
QualiSampler sets
QualiSampler set with LiquiSampler, ViscoSampler, sample bottles (5), a cleaning brush and a carrying case, PTFE
Catalog # 331-2181
Supplier:   BURKLE
QualiSampler sets
Catalog # 331-2181
Supplier:  BURKLE
QualiSampler sets
QualiSampler sets
QualiSampler set with LiquiSampler, ViscoSampler, sample bottles (5), a cleaning brush and a carrying case, PTFE
Price
962,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Complete handy set combining the LiquiSampler and the ViscoSampler with practical accessories.
Delivery information: Set consists of LiquiSampler 100 cm, ViscoSampler 100 cm, case, 5 sample bottles, cleaning brush.
Specifications
  • Description:
    QualiSampler set with LiquiSampler, ViscoSampler, sample bottles (5), a cleaning brush and a carrying case
  • Material:
    PTFE