Order Entry
Austria
Orders LinkContactUsLinkComponent
Stand rods

Stand rods

12 mm, 1250 mm
Catalog # 241-7159
Supplier:   USBECK Laborgeräte
Stand rods
Catalog # 241-7159
Stand rods
Stand rods

12 mm, 1250 mm

Price
25,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel.

  • Robust, very stable
  • Excellent chemical resistance
Specifications
  • Ø:
    12 mm
  • Length:
    1250 mm
  • Seq:
    07
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.