Stand bases
210×130 mm, 1,7 kg
Catalog # 241-7101
Supplier: USBECK Laborgeräte
Jump to:
Product Details & Documents
Stainless steel. Firm, non slip stand for a wide variety of uses. With an M10 threaded hole on face side and adjustable feet for uneven surfaces.
- Rubber feet for a firm footing
- Smooth surfaces for easy, fast cleaning
- Robust
DIN 12892/55
: DIN 12892/55
Specifications
- L×W:210×130 mm
- Seq:02
- Weight:1,7 kg



