Orders LinkContactUsLinkComponent
Forceps
Forceps
Straight, blunt, 115 mm
Catalog # 232-1982
Supplier:   RSG
Forceps
Catalog # 232-1982
Supplier:  RSG
Forceps
Forceps
Straight, blunt, 115 mm
Price
25,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel, PTFE coated.
  • Available in sharp or blunt design
  • Very good chemical and thermal resistance
  • Smooth surfaces prevent contamination and allow easy cleaning
Specifications
  • Type:
    Straight, blunt
  • Length:
    115 mm