Order Entry
Austria
Orders LinkContactUsLinkComponent
 

pH buffer solutions, Specpure®

1,68, ±0,01, HCl/KCl, Colourless, 25 °C, Glass bottle, 4 L
Catalog # 42415.K7
pH buffer solutions, Specpure®
Catalog # 42415.K7
CAS Number:  
pH buffer solutions, Specpure®
pH buffer solutions, Specpure®

1,68, ±0,01, HCl/KCl, Colourless, 25 °C, Glass bottle, 4 L

CAS Number:  
Price
99,30 €
Log in for availability
1 * 4 L
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • pH buffer type:
    HCl/KCl
  • Pk:
    4 L
  • Reference temperature:
    25 °C
  • Colour:
    Colourless
  • pH accuracy:
    ±0,01
  • pH value:
    1,68

Specifications