Order Entry
Austria
Orders LinkContactUsLinkComponent
Spoons, POLY

Spoons, POLY

Straight, 255 mm, 40×20 mm
Catalog # 231-1269
Supplier:   BOCHEM
Spoons, POLY
Catalog # 231-1269
Supplier:  BOCHEM
Spoons, POLY
Spoons, POLY

Straight, 255 mm, 40×20 mm

Price
6,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel 18/10, anti-magnetic. Multipurpose spoon for a wide range of uses. Special shape for light and easy handling.

  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean
Specifications
  • Description:
    Straight
  • Length:
    255 mm
  • Seq:
    07
  • Spoon size:
    40×20 mm