Order Entry
Austria
Orders LinkContactUsLinkComponent
Filter discs

Filter discs

50,8 mm, 1,59 mm
Catalog # 42298.KS
Filter discs
Catalog # 42298.KS
Filter discs

50,8 mm, 1,59 mm

Price
76,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These quartz discs are manufactured by fusing naturally occurring crystalline silica.

  • Ground and polished
Specifications
  • Ø:
    50,8 mm
  • Seq:
    01
  • Thickness:
    1,59 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.