ContactUsLinkComponent
Hose, ptfe, 0.5 x 6 x 7 mm, for the highest standards, good chemical and temperature resistant from -220 ° c to +260 ° c. the ptfe t ube has a low coefficient of friction, is non-stick (easy to clean) and uv-resistant. 1 * 1 m
Catalog # 228-2399
Supplier:  SEMADENI
Hose, ptfe, 0.5 x 6 x 7 mm, for the highest standards, good chemical and temperature resistant from -220 ° c to +260 ° c. the ptfe t ube has a low coefficient of friction, is non-stick (easy to clean) and uv-resistant. 1 * 1 m
Catalog # 228-2399
Supplier:  SEMADENI
Frequently Bought Together