Order Entry
Austria
Orders LinkContactUsLinkComponent
Analytical funnel

Analytical funnel

150 mm, 8 mm, 55 mm
Catalog # 221-1715
Supplier:   Kavalier
Analytical funnel
Catalog # 221-1715
Supplier:  Kavalier
Analytical funnel
Analytical funnel

150 mm, 8 mm, 55 mm

Price
65,30 €
Log in for availability
1 * 1 ST
Product Details & Documents

Borosilicate glass 3.3.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

For rapid filtration.

Specifications
  • Funnel O-Ø:
    55 mm
  • Stem O-Ø:
    8 mm
  • Stem length:
    150 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.