Orders LinkContactUsLinkComponent
Wire baskets, rectangular, with handle
Wire baskets, rectangular, with handle
Without dividers, 480×300×150 mm
Catalog # 216-7513
Supplier:   DELTALAB
Wire baskets, rectangular, with handle
Catalog # 216-7513
Supplier:  DELTALAB
Wire baskets, rectangular, with handle
Without dividers, 480×300×150 mm
Price
370,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Made of stainless steel.
  • Features a strong woven mesh base
  • With folding handles
  • Suitable for bottles and containers
Documents
Specifications
  • Description:
    Without dividers
  • Seq:
    01
  • W×D×H:
    480×300×150 mm