Order Entry
Austria
Orders LinkContactUsLinkComponent
Storage Boxes

Storage Boxes

with hinged lid and 4×6 number of compartments, Transparent, 330×220×53 mm, PS
Catalog # 216-7312
Supplier:   SEMADENI
Storage Boxes
Catalog # 216-7312
Supplier:  SEMADENI
Storage Boxes
Storage Boxes

with hinged lid and 4×6 number of compartments, Transparent, 330×220×53 mm, PS

Price
44,10 €
Log in for availability
1 * 1 ST
Product Details & Documents

These assortment and bearing boxes made of polystyrene (PS) are transparent with hinged lid and multiple longitudinal and transverse divisions.

Specifications
  • Colour:
    Transparent
  • Description:
    with hinged lid and 4×6 number of compartments
  • Material:
    PS
  • Seq:
    02
  • W×D×H:
    330×220×53 mm