Bowls, PP
3000 ml, 100 mm, 240 mm
216-1203
: VITLAB
Bowls, made of white PP.
- Very good chemical resistance
- Very good temperature resistance
- Easy to clean
Ideal for safe storage of food products. With broad, stable edge and circumferential standing ring on the bottom.
- Ext.Ø:240 mm
- Capacity:3000 ml
- Height:100 mm
- Seq:03



