Order Entry
Austria
Orders LinkContactUsLinkComponent
Bottles, narrow neck, rectangular, PVC, with thread

Bottles, narrow neck, rectangular, PVC, with thread

1000 ml, 25 mm, 87,5×61×233 mm
Catalog # 215-3388
Supplier:   Kautex
Bottles, narrow neck, rectangular, PVC, with thread
Catalog # 215-3388
Supplier:  Kautex
Bottles, narrow neck, rectangular, PVC, with thread
Bottles, narrow neck, rectangular, PVC, with thread

1000 ml, 25 mm, 87,5×61×233 mm

Price
3,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

PVC, rectangular, transparent.

  • Crystal clear
  • Secure grip
  • Leakproof
  • Suitable for food
  • Halal certified

Crystal clear presentation of contents, with ribbed sides for secure grip.

Documents

Specifications
  • Thread:
    25 mm
  • Capacity:
    1000 ml
  • Seq:
    03
  • W×D×H:
    87,5×61×233 mm