Order Entry
Austria
Orders LinkContactUsLinkComponent
Bottles, narrow neck, rectangular, PVC, with thread

Bottles, narrow neck, rectangular, PVC, with thread

250 ml, 25 mm, 59×38×151,5 mm
Catalog # 215-3386
Supplier:   Kautex
Bottles, narrow neck, rectangular, PVC, with thread
Catalog # 215-3386
Supplier:  Kautex
Bottles, narrow neck, rectangular, PVC, with thread
Bottles, narrow neck, rectangular, PVC, with thread

250 ml, 25 mm, 59×38×151,5 mm

Price
1,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

PVC, rectangular, transparent.

  • Crystal clear
  • Secure grip
  • Leakproof
  • Suitable for food
  • Halal certified

Crystal clear presentation of contents, with ribbed sides for secure grip.

Documents

Specifications
  • Thread:
    25 mm
  • Capacity:
    250 ml
  • Seq:
    01
  • W×D×H:
    59×38×151,5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.