Assortment Boxes, PP
Assortment box, with hinged lid and 4×8 compartments, Anthracite, 340×260×55 mm
215-0289
: SEMADENI
These assortment boxes, made of PP, have a transparent stable hinged lid with lock and tamper-evident seals.
- Robust and strong design
- All models can be stacked together
- Lid is fixed on each tray so that the contents don't mix even if box is turned upside down
- Colour:Anthracite
- Description:Assortment box, with hinged lid and 4×8 compartments
- Seq:06
- W×D×H:340×260×55 mm
- Cat. No.:215-0289