Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Test tubes

10 ml, 100 mm, 15,8 mm, ungraduated, 13,7 mm
Catalog # 212-3326
Supplier:   DELTALAB
Test tubes
Catalog # 212-3326
Supplier:  DELTALAB
Test tubes
Test tubes

10 ml, 100 mm, 15,8 mm, ungraduated, 13,7 mm

Price
63,30 €
Log in for availability
1 * 500 ST
Product Details & Documents

Polypropylene (PP), ultra clear.

  • Temperature resistant from 0 to 120 °C (short-term up to 130 °C)
  • Centrifugeable up to 15000×g
  • Autoclavable

Precisely shaped reagent and sample tubes for a variety of applications. With round or flat bottom. Unsterile. Graduated.

Specifications
  • Ext.Ø:
    15,8 mm
  • Ø int.:
    13,7 mm
  • Version:
    ungraduated
  • Capacity:
    10 ml
  • Length:
    100 mm
  • Seq:
    08