Test tubes
6 ml, 87,1 mm, 11,9 mm, graduated, 10,3 mm
Supplier: DELTALAB
Jump to:
Product Details & Documents
Polypropylene (PP), ultra clear.
- Temperature resistant from 0 to 120 °C (short-term up to 130 °C)
- Centrifugeable up to 15000×g
- Autoclavable
Precisely shaped reagent and sample tubes for a variety of applications. With round or flat bottom. Unsterile. Graduated.
Specifications
- Ext.Ø:11,9 mm
- Ø int.:10,3 mm
- Version:graduated
- Capacity:6 ml
- Length:87,1 mm
- Seq:05



