Order Entry
Austria
Orders LinkContactUsLinkComponent
pH buffer solutions, Specpure®

pH buffer solutions, Specpure®

7,00, ±0,01, Phosphate, Green, 25 °C, Glass bottle, 500 mL
Catalog # 38711.AP
pH buffer solutions, Specpure®
Catalog # 38711.AP
CAS Number:  
pH buffer solutions, Specpure®
pH buffer solutions, Specpure®

7,00, ±0,01, Phosphate, Green, 25 °C, Glass bottle, 500 mL

CAS Number:  
Price
36,20 €
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • pH buffer type:
    Phosphate
  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • Colour:
    Green
  • pH accuracy:
    ±0,01
  • pH value:
    7,00

Specifications