Order Entry
Austria
Orders LinkContactUsLinkComponent
pH buffer solutions, Specpure®

pH buffer solutions, Specpure®

10,00, ±0,01, Carbonate, Blue, 25 °C, Plastic bottle, 1 L
Catalog # 38709.K2
pH buffer solutions, Specpure®
Catalog # 38709.K2
CAS Number:  
pH buffer solutions, Specpure®
pH buffer solutions, Specpure®

10,00, ±0,01, Carbonate, Blue, 25 °C, Plastic bottle, 1 L

CAS Number:  
Price
55,50 €
Log in for availability
1 * 1 L
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Carbonate
  • Pk:
    1 L
  • Reference temperature:
    25 °C
  • Colour:
    Blue
  • pH accuracy:
    ±0,01
  • pH value:
    10,00

Specifications