Order Entry
Austria
Orders LinkContactUsLinkComponent
pH buffer solutions, Specpure®

pH buffer solutions, Specpure®

10,00, ±0,01, Carbonate, Blue, 25 °C, Glass bottle, 500 mL
Catalog # 38709.AP
pH buffer solutions, Specpure®
Catalog # 38709.AP
CAS Number:  
pH buffer solutions, Specpure®
pH buffer solutions, Specpure®

10,00, ±0,01, Carbonate, Blue, 25 °C, Glass bottle, 500 mL

CAS Number:  
Price
35,30 €
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • pH buffer type:
    Carbonate
  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • Colour:
    Blue
  • pH accuracy:
    ±0,01
  • pH value:
    10,00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.