L(+)-Ascorbic acid 99+% ACS
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
L(+)-Ascorbic acid 99+% ACS
Catalog # 36237.36
Supplier: Thermo Fisher Scientific
CAS Number: 50-81-7
Jump to:
Product Details & Documents
Purity: 99+% CAS No.: 50-81-7 Molecular Formula: C6H8O6 Formula Weight: 176.12 Physical Form: Powder Density: 1.65 Melting Point: ca 191° dec. Beilstein Reference: 84272 Merck Reference: 14,830 MDL No.: MFCD00064328
Synonyms: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
- CAS Number:50-81-7
- Formula:C₆H₈O₆
- MDL Number:MFCD00064328
- Molecular Weight:176.13 g/mol
- Merck Index:14,830
- Boiling Pt:553 °C (1013 hPa)
- Melting Pt:∼ 191 °C (dec.)
- Density:1.65
- Storage Temperature:Ambient
- EINECS:200-066-2
- REACH:exempt
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



