Order Entry
Austria
Orders LinkContactUsLinkComponent
Rods, ceramic, round, solid, AL23

Rods, ceramic, round, solid, AL23

5,0 mm, 600 mm
Catalog # 32931.EH
Rods, ceramic, round, solid, AL23
Catalog # 32931.EH
Rods, ceramic, round, solid, AL23

5,0 mm, 600 mm

Price
138,00 €
Log in for availability
1 * 600 mm
Product Details & Documents

Pure Al₂O₃ Aluminium oxide, AL23.

Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.

Specifications
  • Ext.Ø:
    5,0 mm
  • Length:
    600 mm
  • Seq:
    45
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.