Order Entry
Austria
Orders LinkContactUsLinkComponent
Methylcyclohexane ≥99%

Methylcyclohexane ≥99%

Glass bottle for solids, 500 g
Catalog # 31861.36
Flammable
Health
Irritant
Environment
Methylcyclohexane ≥99%
Catalog # 31861.36
CAS Number:  108-87-2
Methylcyclohexane ≥99%
Methylcyclohexane ≥99%

Glass bottle for solids, 500 g

CAS Number:  108-87-2
Price
135,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99+% CAS No.: 108-87-2 Molecular Formula: C7H14 Formula Weight: 98.19 Flash Point: -4°(-25°F) Physical Form: Liquid Density: 0.771 Refractive Index: 1.4230 Appearance: Colourless Melting Point: -126° Boiling Point: 101° Beilstein Reference: 505972 Merck Reference: 14,6047 UN No.: 2296 MDL No.: MFCD00001497

  • CAS Number:108-87-2
  • Formula:C₆H₁₁CH₃
  • MDL Number:MFCD00001497
  • Molecular Weight:98.19 g/mol
  • Merck Index:14,6047
  • Boiling Pt:101 °C
  • Melting Pt:–126 °C
  • Density:0.771
  • Flash Pt:–4 °C (–25 °F)
  • Storage Temperature:Ambient
  • EINECS:203-624-3
  • UN:2296
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.